Learn More
IKA™ H 99 Protective Cover
Shop All IKA Products
Click to view available options
Temperature (Metric):
Up to 135°C
Unit Size:
Each
Description
- For RET series and RCT Basic magnetic stirrers
- Form-fitting, in-use cover
- Resistant to most acids, alkaline solutions, and organic solvents
- Silicone construction
Specifications
Specifications
| For Use With (Equipment) | RET series and RCT Basic magnetic stirrer |
| Model | H 99 |
| Temperature (Metric) | Up to 135°C |
| Type | In-use Cover |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering