missing translation for 'onlineSavingsMsg'
Learn More

GW7647, MedChemExpress

Product Code. 16433396
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16433396

Brand: Medchem Express HY13861

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GW7647 is a potent PPARα agonist, with EC50s of 6 nM, 1.1 μM, and 6.2 μM for human PPARα, PPARγ and PPARδ, respectively.

GW7647 is a potent PPARα agonist, with EC50s of 6 nM, 1.1 μM, and 6.2 μM for human PPARα, PPARγ and PPARδ, respectively.

Specifications

Chemical Name or Material GW7647
Physical Form Solid
CAS 265129-71-3
Color White
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C29H46N2O3S
For Use With (Application) COVID-19-anti-virus
Solubility Information DMSO : ≥ 60 mg/mL (119.34 mM)
SMILES CC(C)(SC1=CC=C(CCN(C(NC2CCCCC2)=O)CCCCC3CCCCC3)C=C1)C(O)=O
Molecular Weight (g/mol) 502.75
Formula Weight 502.75
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.