Learn More
GW7604, MedChemExpress
Product Code. 16457617
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16457617 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16457617 Supplier Medchem Express Supplier No. HY117153
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
MedChemExpress GW7604 is an antiestrogen. GW7604 is the metabolite of GW5638, which is a high affinity estrogen receptor (ER) antagonist.
GW7604 is an antiestrogen. GW7604 is the metabolite of GW5638, which is a high affinity estrogen receptor (ER) antagonist.
Specifications
Chemical Name or Material GW7604
CAS 195611-82-6
Color White
Purity Grade Notes Research
Molecular Formula C25H22O3
Quantity 5 mg
SMILES O=C(O)/C=C/C1=CC=C(/C(C2=CC=C(O)C=C2)=C(C3=CC=CC=C3)/CC)C=C1
Molecular Weight (g/mol) 370.44
Formula Weight 370.44
Percent Purity 95.73%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death