missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GW 788388

Product Code. 18004322
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18004322

Brand: Tocris Bioscience™ 3264/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective inhibitor of TGF-βRI

Potent inhibitor of transforming growth factor-β type I receptor (ALK5) (IC50 values are 18 and 93 nM for ALK5 binding and for TGF-β cellular assay respectively). Inhibits esophageal squamous cell carcinoma (ESCC)-induced neoangiogenesis. Orally active.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 452342-67-5
Molecular Formula C25H23N5O2

Specifications

Quantity 10 mg
Formula Weight 425.48
Purity >98%
Molecular Weight (g/mol) 425.49
Chemical Name or Material GW 788388
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.