Learn More
GW 5074, MedChemExpress
Product Code. 16480387
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16480387
Brand: Medchem Express HY10542
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress GW 5074 is a potent and selective c-Raf inhibitor with IC50 of 9 nM, and has no effect on the activities of JNK1/2/3, MEK1, MKK6/7, CDK1/2, c-Src, p38 MAP, VEGFR2 or c-Fms.
GW 5074 is a potent and selective c-Raf inhibitor with IC50 of 9 nM, and has no effect on the activities of JNK1/2/3, MEK1, MKK6/7, CDK1/2, c-Src, p38 MAP, VEGFR2 or c-Fms.
Specifications
Chemical Name or Material GW 5074
CAS 220904-83-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C15H8Br2INO2
Quantity 50 mg
Solubility Information DMSO : ≥ 100 mg/mL (191.96 mM)
SMILES O=C1NC2=C(/C1=C/C3=CC(Br)=C(C(Br)=C3)O)C=C(C=C2)I
Molecular Weight (g/mol) 520.94
Formula Weight 520.94
Percent Purity 98.64%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease