Learn More
GW 4064, TRC
Specifications
Specifications
| Chemical Name or Material | GW 4064 |
| CAS | 278779-30-9 |
| Molecular Formula | C28 H22 Cl3 N O4 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C28H22Cl3NO4/c1-16(2)27-21(26(32-36-27)25-22(29)7-4-8-23(25)30)15-35-20-12-11-18(24(31)14-20)10-9-17-5-3-6-19(13-17)28(33)34/h3-14,16H,15H2,1-2H3,(H,33,34)/b10-9+ |
| SMILES | CC(C)c1onc(c1COc2ccc(\C=C\c3cccc(c3)C(=O)O)c(Cl)c2)c4c(Cl)cccc4Cl |
| IUPAC Name | 3-[(E)-2-[2-chloro-4-[[3-(2,6-dichlorophenyl)-5-propan-2-yl-1,2-oxazol-4-yl]methoxy]phenyl]ethenyl]benzoic acid |
| Molecular Weight (g/mol) | 542.84 |
| Formula Weight | 541.0614 |