missing translation for 'onlineSavingsMsg'
Learn More

GW 4064, MedChemExpress

Product Code. p-200095974
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498058 10 mg 10mg
16487836 100 mg 100mg
16440528 5 mg 5mg
16411316 200 mg 200mg
16403987 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16498058 Supplier Medchem Express Supplier No. HY50108

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GW 4064 is a potent FXR agonist with an EC50 of 65 nM.

GW 4064 is a potent FXR agonist with an EC50 of 65 nM.

Specifications

Chemical Name or Material GW 4064
CAS 278779-30-9
Color White
Purity Grade Notes Research
Molecular Formula C28H22Cl3NO4
Quantity 10 mg
Solubility Information DMSO : 125 mg/mL (230.27 mM; Need ultrasonic) ∣DMF : 100 mg/mL (184.22 mM; Need ultrasonic)
SMILES O=C(C1=CC=CC(/C=C/C2=CC=C(C=C2Cl)OCC3=C(ON=C3C4=C(Cl)C=CC=C4Cl)C(C)C)=C1)O
Molecular Weight (g/mol) 542.84
Formula Weight 542.84
Percent Purity 98.02%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.