missing translation for 'onlineSavingsMsg'
Learn More

Guanfacine hydrochloride, MedChemExpress

Product Code. 16443347
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16443347

Brand: Medchem Express HY17416

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Guanfacine hydrochloride, an anti-hypertensive agent, is a selective α2A-adrenoceptor agonist with Kd of 31 nM and displays 60-fold selectivity over α2B-adrenoceptors.

Guanfacine hydrochloride, an anti-hypertensive agent, is a selective α2A-adrenoceptor agonist with Kd of 31 nM and displays 60-fold selectivity over α2B-adrenoceptors.

Specifications

Chemical Name or Material Guanfacine hydrochloride
Physical Form Solid
CAS 29110-48-3
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C9H10Cl3N3O
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 30 mg/mL (106.18 mM) ∣H2O : 20 mg/mL (70.78 mM; Need ultrasonic)
SMILES O=C(NC(N)=N)CC1=C(Cl)C=CC=C1Cl.[H]Cl
Molecular Weight (g/mol) 282.55
Formula Weight 282.55
Percent Purity 99.81%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.