Learn More
Guaiacol-b-D-gentiobioside, TRC
Specifications
Specifications
| Chemical Name or Material | Guaiacol-Beta-D-gentiobioside |
| CAS | 1416253-68-3 |
| Molecular Formula | C19 H28 O12 |
| Quantity | 5 mg |
| Synonym | 2-Methoxyphenyl 6-O-β-D-glucopyranosyl-β-D-glucopyranoside |
| InChI Formula | InChI=1S/C19H28O12/c1-27-8-4-2-3-5-9(8)29-19-17(26)15(24)13(22)11(31-19)7-28-18-16(25)14(23)12(21)10(6-20)30-18/h2-5,10-26H,6-7H2,1H3/t10-,11-,12-,13-,14+,15+,16-,17-,18-,19-/m1/s1 |
| SMILES | COc1ccccc1O[C@@H]2O[C@H](CO[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)[C@@H](O)[C@H](O)[C@H]2O |
| IUPAC Name | (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[[(2R,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(2-methoxyphenoxy)tetrahydropyran-2-yl]methoxy]tetrahydropyran-3,4,5-triol |
| Molecular Weight (g/mol) | 448.42 |
| Formula Weight | 448.1581 |