Learn More
GSTO1 Mouse anti-Human, CoraLite 488, Clone: 5E1E10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-60150-100UL
Product Code. 16800596
  • 450.00 € / 100µL
Please to purchase this item. Need a web account? Register with us today!
Description
Description
The GSTO1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GSTO1. This antibody recognizes human, mouse, rat antigen. The GSTO1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
Specifications
GSTO1
Monoclonal
CoraLite 488
100 μL
9446
IgG1
Immunofluorescence
5E1E10
Mouse
Primary
Human, Rat, Mouse