Learn More
Cytiva GST MultiTrap™ 4B Plates
96-well filter plates for high-throughput screening and parallel purification of GST-tagged proteins
Brand: Cytiva 28-4055-00
Additional Details : Weight : 0.52800kg
Description
- Highly reproducible well-to-well and plate-to-plate results using manual or robotic systems
- High purity in one step with high chemical stability
- Allows loading of unclarified cell lysate samples directly into wells
- Prepacked with Glutathione Sepharose media
Specifications
GST MultiTrap FF and GST MultiTrap 4B are prepacked, 96-well filter plates for reproducible, high-throughput screening and rapid parallel purification of glutathione S-transferase (GST) tagged proteins. | |
4°C to 30°C, 20% Ethanol | |
Filter Plate |
Glutathione Sepharose 4B | |
4 x 96-well Filter Plates |