missing translation for 'onlineSavingsMsg'
Learn More

GSK3787, MedChemExpress

Product Code. 16412737
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16412737

Brand: Medchem Express HY15577

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK3787 is a selective and irreversible peroxisome proliferator-activated receptor δ (PPARδ) antagonist with pIC50 of 6.6.

GSK3787 is a selective and irreversible peroxisome proliferator-activated receptor δ (PPARδ) antagonist with pIC50 of 6.6.

Specifications

Chemical Name or Material GSK3787
Physical Form Solid
CAS 188591-46-0
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C15H12ClF3N2O3S
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 50 mg/mL (127.30 mM)
SMILES O=C(NCCS(=O)(C1=NC=C(C(F)(F)F)C=C1)=O)C2=CC=C(Cl)C=C2
Molecular Weight (g/mol) 392.78
Formula Weight 392.78
Percent Purity 99.04%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.