missing translation for 'onlineSavingsMsg'
Learn More

GSK2256294A, MedChemExpress

Product Code. p-200102168
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16490298

Brand: Medchem Express HY19644

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK2256294A (GSK 2256294) is a selective and orally active inhibitor of soluble epoxide hydrolase (sEH). GSK2256294A inhibits recombinant human sEH, rat sEH orthologs and murine sEH orthologs with IC50s of 27, 61 and 189 pM, respectively. GSK2256294A can be used for the research of chronic obstructive pulmonary disease (COPD) and cardiovascular disease.

GSK2256294A (GSK 2256294) is a selective and orally active inhibitor of soluble epoxide hydrolase (sEH). GSK2256294A inhibits recombinant human sEH, rat sEH orthologs and murine sEH orthologs with IC50s of 27, 61 and 189 pM, respectively. GSK2256294A can be used for the research of chronic obstructive pulmonary disease (COPD) and cardiovascular disease.

Specifications

Chemical Name or Material GSK2256294A
Physical Form Powder
CAS 1142090-23-0
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C21H24F3N7O
For Use With (Application) COVID-19-immunoregulation
Synonym GSK 2256294
Solubility Information DMSO : ≥ 47 mg/mL (105.04 mM)
SMILES O=C([C@H]1C[C@@H](NC2=NC(NC)=NC(C)=N2)CCC1)NCC3=CC=C(C#N)C=C3C(F)(F)F
Molecular Weight (g/mol) 447.46
Formula Weight 447.46
Percent Purity 98.16%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.