missing translation for 'onlineSavingsMsg'
Learn More

GSK189254A, MedChemExpress

Product Code. 16426396
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16426396 100 mg 100mg
16495958 10 mg 10mg
16482107 50 mg 50mg
16458587 5 mg 5mg
16401399 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16426396 Supplier Medchem Express Supplier No. HY14111

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK189254A (GSK189254) is a novel, potent and selective histamine H3 receptor antagonist with pKi values of 9.59-9.90 and 8.51-9.17 for human and rat H3, respectively.

GSK189254A (GSK189254) is a novel, potent and selective histamine H3 receptor antagonist with pKi values of 9.59-9.90 and 8.51-9.17 for human and rat H3, respectively.

Specifications

Chemical Name or Material GSK189254A
CAS 720690-73-3
Color off White
Purity Grade Notes Research
Molecular Formula C21H25N3O2
Quantity 100 mg
Synonym GSK189254
Solubility Information DMSO : 25 mg/mL (71.14 mM; Need ultrasonic)
SMILES O=C(NC)C(C=N1)=CC=C1OC2=CC(CC3)=C(C=C2)CCN3C4CCC4
Molecular Weight (g/mol) 351.44
Formula Weight 351.44
Percent Purity 98.09%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.