Learn More
GSK189254A, MedChemExpress
Description
GSK189254A (GSK189254) is a novel, potent and selective histamine H3 receptor antagonist with pKi values of 9.59-9.90 and 8.51-9.17 for human and rat H3, respectively.
Specifications
Specifications
| Chemical Name or Material | GSK189254A |
| CAS | 720690-73-3 |
| Color | off White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H25N3O2 |
| Quantity | 10 mg |
| Synonym | GSK189254 |
| Solubility Information | DMSO : 25 mg/mL (71.14 mM; Need ultrasonic) |
| SMILES | O=C(NC)C(C=N1)=CC=C1OC2=CC(CC3)=C(C=C2)CCN3C4CCC4 |
| Molecular Weight (g/mol) | 351.44 |
| Show More |