missing translation for 'onlineSavingsMsg'
Learn More

GSK1292263, MedChemExpress

Product Code. 16431217
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16431217

Brand: Medchem Express HY12066

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK-1292263 is an orally available GPR119 agonist with pEC50s of 6.9 and 6.7 for human and rat GPR119, respectively. GSK-1292263 can be used for the research of type 2 diabetes mellitus (T2DM).

GSK-1292263 is an orally available GPR119 agonist with pEC50s of 6.9 and 6.7 for human and rat GPR119, respectively. GSK-1292263 can be used for the research of type 2 diabetes mellitus (T2DM).

Specifications

Chemical Name or Material GSK1292263
CAS 1032823-75-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H28N4O4S
Quantity 50 mg
Solubility Information DMSO : ≥ 20 mg/mL (43.81 mM)
SMILES CC(C1=NOC(N2CCC(CC2)COC3=CN=C(C=C3)C4=CC=C(C=C4)S(C)(=O)=O)=N1)C
Molecular Weight (g/mol) 456.56
Formula Weight 456.56
Percent Purity 98.68%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.