missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSK1292263, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
GSK-1292263 is an orally available GPR119 agonist with pEC50s of 6.9 and 6.7 for human and rat GPR119, respectively. GSK-1292263 can be used for the research of type 2 diabetes mellitus (T2DM).
Specifications
Specifications
| Chemical Name or Material | GSK1292263 |
| CAS | 1032823-75-8 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C23H28N4O4S |
| Quantity | 100 mg |
| Solubility Information | DMSO : ≥ 20 mg/mL (43.81 mM) |
| SMILES | CC(C1=NOC(N2CCC(CC2)COC3=CN=C(C=C3)C4=CC=C(C=C4)S(C)(=O)=O)=N1)C |
| Molecular Weight (g/mol) | 456.56 |
| Formula Weight | 456.56 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction