missing translation for 'onlineSavingsMsg'
Learn More

GSK1059615, MedChemExpress

Product Code. 16464938
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16464938 10 mg 10mg
16487677 5 mg 5mg
16445716 100 mg 100mg
16431187 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16464938 Supplier Medchem Express Supplier No. HY12036

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK1059615 is a dual inhibitor of PI3Kα/β/δ/γ (reversible) and mTOR with IC50 of 0.4 nM/0.6 nM/2 nM/5 nM and 12 nM, respectively.

GSK1059615 is a dual inhibitor of PI3Kα/β/δ/γ (reversible) and mTOR with IC50 of 0.4 nM/0.6 nM/2 nM/5 nM and 12 nM, respectively.

Specifications

Chemical Name or Material GSK1059615
Physical Form Solid
CAS 958852-01-2
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C18H11N3O2S
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 5 mg/mL (15.00 mM; Need ultrasonic)
SMILES O=C(S/C1=C\C2=CC=C3N=CC=C(C3=C2)C4=CC=NC=C4)NC1=O
Molecular Weight (g/mol) 333.36
Formula Weight 333.36
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.