Learn More
GSK-843, MedChemExpress
Product Code. 16437967
Encompass_Preferred
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Product Code. 16437967
Brand: Medchem Express HY125402
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress GSK-843 (GSK'843) is a receptor-interacting protein kinase 3 (RIP3 or RIPK3) inhibitor, which binds RIP3 kinase domain with an IC50 of 8.6 nM, and inhibits kinase activity with an IC50 of 6.5 nM. GSK-843 can be used for the research of inflammation.
GSK-843 (GSK'843) is a receptor-interacting protein kinase 3 (RIP3 or RIPK3) inhibitor, which binds RIP3 kinase domain with an IC50 of 8.6 nM, and inhibits kinase activity with an IC50 of 6.5 nM. GSK-843 can be used for the research of inflammation.
Specifications
Chemical Name or Material GSK-843
CAS 1601496-05-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C19H15N5S2
Quantity 5 mg
Synonym GSK'843
Solubility Information DMSO : 50 mg/mL (132.45 mM; Need ultrasonic)
SMILES NC1=NC=C(C2=CC(C)=NN2C)C3=C1C(C4=CC=C(SC=N5)C5=C4)=CS3
Molecular Weight (g/mol) 377.49
Formula Weight 377.49
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation