Learn More
GSK 650394, MedChemExpress
Product Code. 16472467
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16472467 50 mg 50mg
16436358 10 mg 10mg
16428947 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16472467 Supplier Medchem Express Supplier No. HY15192
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress GSK 650394 is a novel SGK inhibitor with IC50 of 62 nM and 103 nM for SGK1 and SGK2 in the SPA assay respectively. GSK 650394 also inhibits influenza virus replication.
GSK 650394 is a novel SGK inhibitor with IC50 of 62 nM and 103 nM for SGK1 and SGK2 in the SPA assay respectively. GSK 650394 also inhibits influenza virus replication.
Specifications
Chemical Name or Material GSK 650394
CAS 890842-28-1
Color Light Nude
Purity Grade Notes Research
Molecular Formula C25H22N2O2
Quantity 50 mg
Solubility Information DMSO : ≥ 40.7 mg/mL (106.42 mM)
SMILES O=C(O)C(C=C1)=C(C2CCCC2)C=C1C3=CNC(C3=C4)=NC=C4C5=CC=CC=C5
Molecular Weight (g/mol) 382.45
Formula Weight 382.45
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease