missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GSDMC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101660
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GSDMC.
The N-terminal moiety promotes pyroptosis. May be acting by homooligomerizing within the membrane and forming pores (PubMed:27281216). The physiological relevance of this observation is unknown (Probable). [UniProt].
Specifications
| GSDMC | |
| Polyclonal | |
| Unconjugated | |
| GSDMC | |
| gasdermin C; gasdermin C1; gasdermin-C; Gasdermin-C1; Gsdmc; Gsdmc1; melanoma-derived leucine zipper, extra-nuclear factor; melanoma-derived leucine zipper-containing extranuclear factor; MLZE; mMLZE | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 56169 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9BYG8 | |
| GSDMC | |
| A synthesized peptide derived from human GSDMC(Accession Q9BYG8), corresponding to amino acid residues E108-V158. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction