missing translation for 'onlineSavingsMsg'
Learn More
Learn More
- Home
- Products
- Personal Protective Equipment
- Gloves
- Dry Box Gloves
- AL Glove Systems GSB Isolator Box Gloves
AL Glove Systems GSB Isolator Box Gloves
Product Code. 31337486
Change view
Click to view available options
Size:
Large, 9.5
Medium, 8
X-Large, 11
Length (Metric):
800 mm
920 mm
Thickness (Metric):
0.4 mm
0.6 mm
Diameter (Metric) Cuff:
160 mm
180 mm
200 mm
225 mm
250 mm
300 mm
34 product options available for selection
| Product Code. | Size | Length (Metric) | Thickness (Metric) | Diameter (Metric) Cuff |
|---|---|---|---|---|
| 31337486 | X-Large, 11 | 800 mm | 0.6 mm | 225 mm |
| 31337400 | Medium, 8 | 800 mm | 0.4 mm | 180 mm |
| 31337436 | Medium, 8 | 800 mm | 0.6 mm | 180 mm |
| 31337484 | Medium, 8 | 800 mm | 0.4 mm | 200 mm |
| 31337398 | Medium, 8 | 800 mm | 0.6 mm | 200 mm |
| 31337454 | Medium, 8 | 800 mm | 0.4 mm | 225 mm |
| 31337483 | Medium, 8 | 800 mm | 0.6 mm | 225 mm |
| 31337482 | Large, 9.5 | 800 mm | 0.4 mm | 160 mm |
| 31337497 | Large, 9.5 | 800 mm | 0.6 mm | 160 mm |
| 31337429 | Large, 9.5 | 800 mm | 0.4 mm | 180 mm |
| 31337439 | Large, 9.5 | 800 mm | 0.6 mm | 200 mm |
| 31337416 | Large, 9.5 | 800 mm | 0.4 mm | 225 mm |
| 31337440 | Large, 9.5 | 800 mm | 0.6 mm | 225 mm |
| 31337517 | Large, 9.5 | 800 mm | 0.4 mm | 250 mm |
| 31337495 | Large, 9.5 | 800 mm | 0.6 mm | 250 mm |
| 31337424 | Large, 9.5 | 800 mm | 0.4 mm | 300 mm |
| 31337508 | Large, 9.5 | 800 mm | 0.6 mm | 300 mm |
| 31337461 | Large, 9.5 | 920 mm | 0.4 mm | 200 mm |
| 31337476 | Large, 9.5 | 920 mm | 0.6 mm | 200 mm |
| 31337494 | Large, 9.5 | 920 mm | 0.4 mm | 250 mm |
| 31337469 | Large, 9.5 | 920 mm | 0.6 mm | 250 mm |
| 31337473 | X-Large, 11 | 800 mm | 0.4 mm | 180 mm |
| 31337396 | X-Large, 11 | 800 mm | 0.6 mm | 200 mm |
| 31337407 | X-Large, 11 | 800 mm | 0.4 mm | 225 mm |
| 31337493 | X-Large, 11 | 800 mm | 0.4 mm | 250 mm |
| 31337419 | X-Large, 11 | 800 mm | 0.6 mm | 250 mm |
| 31337401 | X-Large, 11 | 800 mm | 0.4 mm | 300 mm |
| 31337457 | X-Large, 11 | 920 mm | 0.4 mm | 300 mm |
| 31337518 | X-Large, 11 | 800 mm | 0.6 mm | 300 mm |
| 31337399 | X-Large, 11 | 920 mm | 0.4 mm | 225 mm |
| 31337467 | X-Large, 11 | 920 mm | 0.6 mm | 225 mm |
| 31337447 | X-Large, 11 | 920 mm | 0.4 mm | 250 mm |
| 31337488 | X-Large, 11 | 920 mm | 0.6 mm | 250 mm |
| 31337462 | X-Large, 11 | 920 mm | 0.6 mm | 300 mm |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 34 options available.
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
AL Glove Systems GSB gloves are made of bromo-butyl rubber (BBR) with high gas impermeability and high resistance to a wide range of polar hydrocarbons such as acids, esters, ketones, and amine derivatives. They are highly flexible with a high sensitivity. The GSB Glove is classified as PAH-free.
- Impermeable to gas
- High resistance to the permeation of mainly oxygen, nitrogen, inert gases, etc. during production under inert gas atmosphere in inert boxes for the production of electronic components and microcomponents
Flexible:
- Offers good sensitivity in handling
- Resistant to chemicals
- High resistance to a wide range of chemicals and toxins
PERMEATION EN ISO 374-1:2016/A1:2018
- A Methanol Protection Level 6
- B Acetone Protection Level 6
- I Ethyl acetate Protection Level 3
- K Sodium hydroxide 40% Protection Level 6
- L Sulphuric acid 96% Protection Level 6
- N Acetic acid 99% Protection Level 6
- O Ammonium hydroxide 25% Protection Level 6
- T Formaldehyde 37% Protection Level 6
Specifications
Specifications
| Certifications/Compliance (Europe) | PPE Category III Directive 2016/425 EC Type |
| Chlorinated | No |
| Color | Black |
| Hand | Ambidextrous |
| Latex Free | Yes |
| Material | Butyl |
| Product Type | Dry Box, Isolator Gloves |
| Size | X-Large, 11 |
| Temperature (English) Max. | 194°F |
| Temperature (Metric) Max | 90°C |
| Thickness (English) | 24 mil |
| Thickness (Metric) | 0.6 mm |
| For Use With (Application) | Handling Sensitive or Hazardous Materials in an Inert or Contained Atmosphere |
| For Use With (Equipment) | Dry Box, Isolator, RABS |
| Grip Design | Smooth |
| Manufacturing/QMS Audit Standards | ISO 9001:2015 |
| Packaging Type | Carton |
| Temperature (English) Min | -40°F |
| Temperature (Metric) Min. | -40°C |
| Length (English) | 32 in. |
| Length (Metric) | 800 mm |
| Diameter (English) Cuff | 9 In. |
| Diameter (Metric) Cuff | 225 mm |
| Show More Show Less |
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?