missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRK6 Polyclonal Antibody

Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
GRK6 is a member of the guanine nucleotide-binding protein coupled receptor kinase subfamily of the serine/threonine protein kinase family. GRK6 phosphorylates the activated forms of G protein-coupled receptors to initiate their deactivation. Several transcript variants encoding different isoforms have been described for GRK6.
Specifications
Specifications
| Antigen | GRK6 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Grk6 |
| Gene Accession No. | O70293, P43250, P97711 |
| Gene Alias | activin receptor IIA; activin receptor type IIA; Activin receptor type-2A; ActrIIa; ACTR-IIA; Acvr2; Acvr2a; EC 2.7.11.16; G protein-coupled receptor kinase 6; g protein-coupled receptor kinase GRK6; Gprk6; G-protein coupled receptor kinase 6; GRK6; kinase GRK6; TactrII |
| Gene Symbols | Grk6 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?