missing translation for 'onlineSavingsMsg'
Learn More

Carl Roth™ Green Labels

Product Code. 10105003
Encompass
Change view
Click to view available options
For Use With (Equipment):
0.5 mL vessel
1.5 to 2 mL vessel
1.5/2 mL vessels
Unit Size:
1 roll
Pack of 1000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10105003 0.5 mL vessel 1 roll
10290042 1.5/2 mL vessels Pack of 1000
10676603 1.5 to 2 mL vessel -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10105003 Supplier Carl Roth™ Supplier No. X613.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Color Green
Product Type Label
For Use With (Application) Without drying out and falling off sticking on any plastic, z. B. during steam sterilization, in boiling water baths, in refrigerators, in agitation and shakers, centrifuges etc.
For Use With (Equipment) 0.5 mL vessel
Temperature Range (Metric) -96 to 80°C
Diameter (Metric) 9 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.