Learn More
Tenak™ Green Cardboard Cryobox
Product Code. 15270604 Shop All TENAK Products
Change view
Click to view available options
Dimensions (L x W x H):
133 x 133 x 32 mm
133 x 133 x 50 mm
133 x 133 x 75 mm
136 x 136 x 32 mm
136 x 136 x 50 mm
133 x 133 x 100 mm
133 x 133 x 130 mm
136 x 136 x 100 mm
136 x 136 x 130 mm
Unit Size:
Pack of 36
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) unitSize
15270604 136 x 136 x 100 mm Pack of 36
15290534 133 x 133 x 100 mm Pack of 36
15220614 136 x 136 x 130 mm Pack of 36
15280524 133 x 133 x 130 mm Pack of 36
11577294 133 x 133 x 32 mm Pack of 36
15230594 136 x 136 x 32 mm Pack of 36
15190877 133 x 133 x 50 mm Pack of 36
15280594 136 x 136 x 50 mm Pack of 36
15250524 133 x 133 x 75 mm Pack of 36
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 15270604 Supplier Tenak™ Supplier No. TE22543
Please to purchase this item. Need a web account? Register with us today!
Cryobox made of water-repellent cardboard for long-lasting durability.
  • Constructed of fibreboard/cardboard
  • Water-repellent for longer durability
  • Can be used with a large range of grid dividers
Specifications
Color Green
Material Cardboard
Shape Square
Type Cryo Box
Dimensions (L x W x H) 136 x 136 x 100 mm