Learn More
MULTIROIR Gray Euro-Standard Full Container
Product Code. p-7217334
Click to view available options
Dimensions (L x W x H):
400 x 300 x 80 mm
600 x 400 x 80 mm
300 x 200 x 190 mm
400 x 300 x 120 mm
400 x 300 x 290 mm
400 x 300 x 320 mm
800 x 600 x 120 mm
800 x 600 x 230 mm
800 x 600 x 340 mm
800 x 600 x 450 mm
Unit Size:
Each
Product Code. 17276910
Brand: MULTIROIR SPK3017_GREY
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The Euro standard plastic storage bin is compatible with all containers meeting European standards and in particular Euro pallets
  • The Euro standard plastic storage bin is compatible with all containers meeting European standards and in particular Euro pallets
  • The Euro standard container from Multiroir allows the storage, transport and supply of goods in manual or automated mode, indoors or outdoors
  • Manufactured in thick polypropylene and therefore undeformable and very robust
  • Standard silver gray color
  • Euro container with full walls
  • Resistant to acids and bases, cold and heat
  • Food grade
  • Ergonomic handles for optimal handling
  • Smooth walls for easy cleaning
Specifications
Type Storage Box
Material Polypropylene
Color Gray
Shape Rectangular
Includes Closed handles
Dimensions (L x W x H) 300 x 200 x 190 mm