missing translation for 'onlineSavingsMsg'
Learn More

Graphite electrode, crater-drillpoint/undercut, 4.57mm dia, 38.10mm length, volume 0.040cc, 99.9995% (metals basis), Thermo Scientific™

Product Code. 15423735
Change view
Click to view available options
Quantity:
100 Pc.
Packaging:
Tube
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15423735 100 Pc. Tube Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15423735 Supplier Thermo Scientific Alfa Aesar Supplier No. 040788.KN
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Used as Homogeneous Catalysts. They are widely used in electric arc furnaces (EAF) and ladle furnaces (LF) for the melting of steel, non-ferrous metals and some non-metallic minerals.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Graphite electrode
Packaging Tube
Quantity 100 Pc.
Solubility Information Insoluble in water.
Percent Purity 99.9995%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.