missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Granulins Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527275
This item is not returnable.
View return policy
Description
Recommended positive controls: HepG2, mouse kidney, rat kidney. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PCDGF is a glycoprotein and growth factor involved in the regulation of mitosis, early embryonic development, cell division, cell migration, cell survival, and hypothalamic differentiation. PCDGF is also known as progranulin because it is the precursor for smaller proteins known as epithelins or granulins.
Specifications
| Granulins | |
| Polyclonal | |
| Unconjugated | |
| GRN | |
| acrogranin; acrogranulin; CLN11; epithelin; Epithelin precursor; Epithelin/granulin precursor; Epithelin-1; Epithelin-2; GEP; Glycoprotein of 88 Kda; GP88; granulin; Granulin A; Granulin B; Granulin C; Granulin D; Granulin E; Granulin F; Granulin G; Granulin precursor; Granulin-1; Granulin-2; Granulin-3; Granulin-4; Granulin-5; Granulin-6; Granulin-7; granulin-epithelin; granulins; Grn; Paragranulin; PC cell-derived growth factor; PCDGF; PEPI; PGRN; Proepithelin; progranulin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2896 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1.43 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P28799 | |
| GRN | |
| Recombinant protein encompassing a sequence within the center region of human Granulins. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction