Learn More
Invitrogen™ GRAMD1A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552617
Description
Immunogen sequence: SEDEDYVSPL QLNGLGTPKE VGDVIALSDI TSSGAADRSQ EPSPVGSRRG HVTPNLSRAS SDADHGAEED KEEQVDSQPD ASSSQTVTPV AEPPSTEPTQ PDGPTTLGPL DLLPS Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 85%.
Cholesterol transporter that mediates non-vesicular transport of cholesterol from the plasma membrane (PM) to the endoplasmic reticulum (ER). Contains unique domains for binding cholesterol and the PM, thereby serving as a molecular bridge for the transfer of cholesterol from the PM to the ER. Plays a crucial role in cholesterol homeostasis and has the unique ability to localize to the PM based on the level of membrane cholesterol. In lipid-poor conditions localizes to the ER membrane and in response to excess cholesterol in the PM is recruited to the endoplasmic reticulum-plasma membrane contact sites (EPCS) which is mediated by the GRAM domain. At the EPCS, the sterol-binding VASt/ASTER domain binds to the cholesterol in the PM and facilitates its transfer from the PM to ER. May play a role in tumor progression. Plays a role in autophagy regulation and is required for biogenesis of the autophagosome (PubMed:31222192). This function in autophagy requires its cholesterol-transfer activity (PubMed:31222192). [UniProt].
Specifications
| GRAMD1A | |
| Polyclonal | |
| Unconjugated | |
| GRAMD1A | |
| 1300003M23Rik; AI415002; AW547240; D7Bwg0611e; GRAM domain containing 1A; GRAM domain-containing protein 1A; GRAMD1A; KIAA1533; Protein Aster-A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 57655 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96CP6 | |
| GRAMD1A | |
| Recombinant protein corresponding to Human GRAMD1A. Recombinant protein control fragment (Product #RP-90008). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.