Learn More
Hecht Karl™ Graduated Glass Cylinder
3.98 € - 26.34 €
Product Code | Brand | Item Description | Capacity (Metric) | Height (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Item Description | Capacity (Metric) | Height (Metric) | Price | Quantity & Availability | |||
10014111
|
Hecht Karl™
44401042 |
Low Form | 200 mL | 195 mm |
10.92 €
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||
10238391
|
Hecht Karl™
44400030 |
Tall Form | 100 mL | 250 mm |
3.98 €
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||
10410762
|
Hecht Karl™
44401012 |
Low Form | 10 mL | 90 mm |
4.82 €
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||
10450192
|
Hecht Karl™
44400066 |
Tall Form | 1000 mL | 440 mm |
26.34 €
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||
10525762
|
Hecht Karl™
44400048 |
Tall Form | 250 mL | 325 mm |
6.47 €
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||
10746952
|
Hecht Karl™
44400042 |
Tall Form | 200 mL | 265 mm |
6.47 €
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||
Specifications
Graduated Cylinder | |
Glass |