Learn More
Sartorius Stedim Biotech™ Grade 6 S/N Creped Papers for Qualitative and Technical Analyses, Folded
Sartorius Stedim Biotech™ Grade 6 S/N Creped Papers for Qualitative and Technical Analyses, Folded are suitable for analytical and technical purposes and feature a fast flow rate due to their structure.
230.00 € - 418.00 €
Specifications
Basis Weight | 145 g/m2 |
---|---|
Format | Folded |
Material | Cotton Linters, Refined Pulp |
Particle Retention | 6 to 14 μm |
Filtration Speed | 12 sec. |
Product Code | Brand | Diameter (Metric) | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Quantity | Price | Quantity & Availability | ||||
15992990
|
Sartorius Stedim Biotech™
FT-4-314-125 |
125 mm | 100/Pk. |
230.00 €
Pack of 100 |
Estimated Shipment: 03-07-2024
Log in to see stock. |
||||
15903000
|
Sartorius Stedim Biotech™
FT-4-314-185 |
185 mm | 100/Pk. |
235.00 €
Pack of 100 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||||
15913000
|
Sartorius Stedim Biotech™
FT-4-314-240 |
240 mm | 100/Pk. |
249.00 €
Pack of 100 |
Estimated Shipment: 25-06-2024
Log in to see stock. |
||||
15923000
|
Sartorius Stedim Biotech™
FT-4-314-320 |
320 mm | 100/Pk. |
306.00 €
Pack of 100 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||||
15933000
|
Sartorius Stedim Biotech™
FT-4-314-500 |
500 mm | 50/Pk. |
418.00 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
Description
- Made of refined pulp and cotton linters with > 95% alpha-cellulose
- Ash content between 0.1 and 0.15%
- Medium fast filtering
- Wet-strengthened
- Non-sterile, bulk-packed
Specifications
145 g/m2 | |
Cotton Linters, Refined Pulp | |
12 sec. | |
0.55 mm |
Folded | |
6 to 14 μm | |
6 S/N | |
Qualitative Filter Paper |