Learn More
Sartorius™ Grade 1292 Wet-strengthened Folded Filter Qualitative Filter Papers
Used for analytical purposes and routine analyses, whenever no gravimetric analyses are required
Brand: Sartorius™ FT-4-210-240
Additional Details : Weight : 0.10000kg
Description
- Made of refined pulp and cotton linters with an >95% α-cellulose content
- Ash content <0.1%
- Precipates fine crystalline
- Medium dense, medium fast filtering
- Supplied in folded format
Specifications
Folded | |
5 to 8 μm | |
Used for analytical purposes and routine analyses, whenever no gravimetric analyses are required | |
100/Pk. | |
Qualitative Filter Paper | |
240 mm |
Cotton Linters, Refined Pulp | |
50μm/sec. | |
1292 | |
0.17 mm | |
240 mm |