missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPX7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585441
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Positive Control: mouse liver. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Human GPX 7 is believed to be a member of the glutathione peroxidase family of proteins. These proteins play important roles in cellular defense mechanisms against oxidative damage. This is accomplished by catalyzing the reduction of hydroperoxides and hydrogen peroxide through the reducing substrate, glutathione.
Specifications
| GPX7 | |
| Polyclonal | |
| Unconjugated | |
| GPX7 | |
| 3110050F08Rik; AI327032; CL683; FLJ14777; glutathione peroxidase 6; glutathione peroxidase 7; GPX6; Gpx7; GPx-7; GSHPx-7; HGNC:4559; non-selenocysteine containing phospholipid hydroperoxide glutathione peroxidase; NPGPx; UNQ469/PRO828 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 67305 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1.03 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q99LJ6 | |
| GPX7 | |
| The immunogen used to generate this antibody corresponds to mouse m-NPGPX/GPX7. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction