missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPRASP2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15526262
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15526262 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15526262 Supplier Invitrogen™ Supplier No. PA543854

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: EQESLLQPDQ PSPEFTFQYD PSYRSVREIR EHLRARESAE SESWSCSCIQ Sequence homology: Cow: 92%; Dog: 92%; Guinea Pig: 91%; Horse: 92%; Human: 100%; Mouse: 92%; Pig: 100%; Rabbit: 85%; Rat: 92%.

The protein encoded by this gene is a member of a family that regulates the activity of G protein-coupled receptors (GPCRs). The encoded protein has been shown to be capable of interacting with several GPCRs, including the M1 muscarinic acetylcholine receptor and the calcitonin receptor. Several transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPRASP2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GPRASP2
Gene Accession No. Q96D09
Gene Alias 5330440H13Rik; FLJ35662; FLJ37327; G protein-coupled receptor associated sorting protein 2; GASP2; GASP-2; GPRASP2; G-protein coupled receptor-associated sorting protein 2; prion protein ligand 5; Prpl5; RGD1561019
Gene Symbols GPRASP2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human GPRASP2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114928
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.