missing translation for 'onlineSavingsMsg'
Learn More

GPR85 Antibody (1042202) [PerCP], Novus Biologicals™

Product Code. 30678033 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30678033 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30678033 Supplier Novus Biologicals Supplier No. FAB10761C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GPR85 Monoclonal antibody specifically detects GPR85 in Human samples. It is validated for Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR85
Applications Immunocytochemistry
Classification Monoclonal
Clone 1042202
Conjugate PerCP
Formulation PBS
Gene Alias FLJ50993, FLJ51159, FLJ99433, G protein-coupled receptor 85, probable G-protein coupled receptor 85, seven transmembrane helix receptor, SREB, SREB2Super conserved receptor expressed in brain 2
Host Species Mouse
Immunogen Human GPR85 extracellular domain synthetic peptide
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 54329
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.