missing translation for 'onlineSavingsMsg'
Learn More

GPR83 Monoclonal Antibody (OTI3E8), TrueMAB™, OriGene

Product Code. 17506144
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17506144 30 μL 30µL
17586134 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17506144 Supplier Origene Technologies Supplier No. TA811836S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GPR83 Monoclonal antibody specifically detects GPR83 in Human samples. It is validated for Western Blot

Specifications

Antigen GPR83
Applications Western Blot
Classification Monoclonal
Clone OTI3E8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene GPR17
Gene Accession No. Q13304
Gene Alias AW045253; G protein-coupled receptor 72; G protein-coupled receptor 83; Gir; glucocorticoid induced receptor; glucocorticoid-induced receptor; GPR72; Gpr83; G-protein coupled receptor 72; Hcepto9; Jp05; KIAA1540; Probable G-protein coupled receptor 83; RP105; RP39; RP82
Host Species Mouse
Immunogen Synthetic peptide
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2840
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.