missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPR63 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16667424
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16667424 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16667424 Supplier Invitrogen™ Supplier No. PA5100945

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GPR63.

G protein-coupled receptors (GPCRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. GPR63 is a member of this family (subfamily: lysophospholipid/lysosphingolipid) and its expression has been reported in several brain regions, including caudate, frontal cortex, hypothalamus, midbrain, and thalamus, as well as in small intestine, stomach, and thymus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR63
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GPR63
Gene Accession No. Q9BZJ6, Q9EQQ3
Gene Alias brain expressed G-protein-coupled receptor PSP24 beta; G protein-coupled receptor 63; G protein-coupled receptor PSP24-2; Gpr63; Probable G-protein coupled receptor 63; PSP24(beta); PSP24-2; PSP24B; PSP24beta; PSP24-beta
Gene Symbols GPR63
Host Species Rabbit
Immunogen A synthesized peptide derived from human GPR63(Accession Q9BZJ6), corresponding to amino acid residues K196-I246.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81006, 81491
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.