Learn More
Invitrogen™ GPR40 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595511
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human PANC-1 whole cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
This gene encodes a member of the GP40 family of G protein-coupled receptors that are clustered together on chromosome 19. The encoded protein is a receptor for medium and long chain free fatty acids and may be involved in the metabolic regulation of insulin secretion. Polymorphisms in this gene may be associated with type 2 diabetes.
Specifications
| GPR40 | |
| Polyclonal | |
| Unconjugated | |
| FFAR1 | |
| FFA1R; FFAR; Ffar1; Free fatty acid receptor 1; G protein-coupled receptor 40; G protein-coupled receptor GPR40; GPCR40; GPR40; G-protein coupled receptor 40; HGNC:4498 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 233081, 266607, 2864 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| O14842, Q76JU9, Q8K3T4 | |
| FFAR1 | |
| A synthetic peptide corresponding to a sequence of human GPCR GPR40 (YVGCLRALARSGLTHRRKLRAAWVA). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.