missing translation for 'onlineSavingsMsg'
Learn More

GPR31 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15537321
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15537321 100 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15537321 Supplier Invitrogen Supplier No. PA3037

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

GPR31 Polyclonal Antibody

GPR31 Polyclonal Antibody for IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR31
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 0.05% sodium azide
Gene GPR31
Gene Accession No. O00270
Gene Alias 12-(S)-HETE acid receptor; 12-(S)-HETE receptor; 12-(S)-hydroxy-5,8,10,14-eicosatetraenoic acid (HETE) receptor; 12-(S)-hydroxy-5,8,10,14-eicosatetraenoic acid receptor; 12-HETER; bA517H2.2 (G protein-coupled receptor 31); G protein-coupled receptor 31; GPR31; G-protein coupled receptor 31; HETER; HETER1; hydroxyeicosatetraenoic (HETE) acid receptor 1; probable G-protein coupled receptor 31; TCP10C
Gene Symbols GPR31
Host Species Rabbit
Immunogen KLH conjugated Cys-TLRGKGQAAEPPDFNPRDSYS.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2853
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.