missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR146 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-16718
This item is not returnable.
View return policy
Description
GPR146 Polyclonal specifically detects GPR146 in Human samples. It is validated for Western Blot.
Specifications
| GPR146 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0) and 50% Glycerol with 0.01% Thimerosal | |
| G protein-coupled receptor 146, G protein-coupled receptor PGR8, PGR8G-protein coupled receptor PGR8, probable G-protein coupled receptor 146 | |
| Rabbit | |
| 37 kDa | |
| 0.1 mL | |
| GPCR | |
| 115330 | |
| Human | |
| IgG |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q96CH1 | |
| GPR146 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human GPR146. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Expected to cross react based on sequence identity: Monkey (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction