missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPNMB Monoclonal Antibody (HOST5DS), APC, eBioscience™

Product Code. 16516950
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16516950 100 Tests Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16516950 Supplier Invitrogen™ Supplier No. 17983842

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Mouse Monoclonal Antibody

This HOST5DS monoclonal antibody recognizes human GPNMB also known as Osteoactivin, DC-HIL and HGFIN. This type-1 transmembrane glycoprotein was described independently as a bone development factor, tumor related molecule, neurokinin, and immune regulator. Expressed on macrophages and dendritic cells, GPNMB is a ligand for proteoglycan Syndecan-4 (SD-4), which is found on T cells, and is known to inhibit their activation and proliferation. Hence, surface expression of GPNMB by myeloid cells may be indicative of their tolerogenic properties. Various cancers express GPNMB, and it is suspected to help in immune evasion. Treatment with IL-10 and various immunosuppressive drugs (e.g., dexamethasone) upregulate GPNMB on macrophages and dendritic cells. THP-1 and U937 cell lines constitutively express GPNMB. GPNMB may also be secreted. The HOST5DS antibody will work on paraformaldehyde-fixed and permeabilized cells, and can be used for intracellular staining. Please, note that enzymatic tissue dissociation may reduce the amount of surface GPNMB, and thereby negatively impact the brightness of the staining. Applications Reported: This HOST5DS antibody has been reported for use in flow cytometric analysis. Applications Tested: This HOST5DS antibody has been pre-diluted and tested by flow cytometric analysis of normal human blood monocyte-derived macrophages. This may be used at 5 μL (0.125 μg) per test. A test is defined as the amount ( μg) of antibody...

The protein encoded by this gene is a type I transmembrane glycoprotein which shows homology to the pMEL17 precursor, a melanocyte-specific protein. GPNMB shows expression in the lowly metastatic human melanoma cell lines and xenografts but does not show expression in the highly metastatic cell lines. GPNMB may be involved in growth delay and reduction of metastatic potential. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPNMB
Applications Flow Cytometry
Classification Monoclonal
Clone HOST5DS
Concentration 5 μL/Test
Conjugate APC
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene GPNMB
Gene Accession No. Q14956
Gene Alias Dchil; DC-HIL; dendritic cell-associated heparin sulfate proteoglycan-dependent integrin ligand; dendritic cell-associated transmembrane protein; glycoprotein (transmembrane) nmb; glycoprotein nmb; glycoprotein nmb-like protein; glycoprotein nonmetastatic melanoma protein B; Gpnmb; hematopoietic growth factor inducible neurokinin-1; Hematopoietic growth factor inducible neurokinin-1 type; Hgfin; ipd; iris pigment dispersion; NMB; Osteoactivin; si:ch211-155k24.6; transmembrane glycoprotein HGFIN; transmembrane glycoprotein NMB; Unknown (protein for MGC:133733); UNQ1725/PRO9925
Gene Symbols GPNMB
Host Species Mouse
Immunogen FC-tagged full protein expressed in insect cells.
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10457
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.