Learn More
Abnova™ GPC1/CEN2q FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Brand: Abnova™ FG0198.200uL
Additional Details : Weight : 0.02000kg
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
FLJ38078,glypican | |
2817 | |
200 μL |
Tube | |
glypican 1 | |
GPC1 | |
DNA/RNA | |
Liquid |