Learn More
GP531, MedChemExpress
Product Code. 16412378
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16412378 5 mg 5mg
16410179 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16412378 Supplier Medchem Express Supplier No. HYU00116
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress GP531 is a potent, second-generation adenosine regulating agent, is pharmacologically silent under basal conditions but increases localized endogenous adenosine during ischemia.
GP531 is a potent, second-generation adenosine regulating agent, is pharmacologically silent under basal conditions but increases localized endogenous adenosine during ischemia.
Specifications
Chemical Name or Material GP531
CAS 142344-87-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H21N5O4
Quantity 5 mg
Solubility Information DMSO : 125 mg/mL (359.85 mM; Need ultrasonic)
SMILES O=C(C1=C(N)N([C@H]2[C@@H]([C@@H]([C@@H](CNCC3=CC=CC=C3)O2)O)O)C=N1)N
Molecular Weight (g/mol) 347.37
Formula Weight 347.37
Percent Purity 98.95%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation