missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goosecoid Antibody (OTI1D7), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72409MFV500
This item is not returnable.
View return policy
Description
Goosecoid Monoclonal antibody specifically detects Goosecoid in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
| Goosecoid | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry | |
| OTI1D7 | |
| Flow Cytometry | |
| goosecoid homeobox, homeobox protein goosecoid | |
| Human recombinant protein fragment corresponding to amino acids 107-257 of human GSC (NP_776248) produced in E.coli. | |
| 0.1 mL | |
| Stem Cells | |
| 145258 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction