missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gonadotropin Inducible Transcription Repressor 1 Antibody (4G11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00092283-M01
This item is not returnable.
View return policy
Description
Gonadotropin Inducible Transcription Repressor 1 Monoclonal antibody specifically detects Gonadotropin Inducible Transcription Repressor 1 in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
Specifications
| Gonadotropin Inducible Transcription Repressor 1 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| zinc finger protein 213, Zinc finger protein with KRAB and SCAN domains 21, ZKSCAN21CR53Putative transcription factor CR53 | |
| GIOT-1 (NP_694989.2, 121 a.a. ∽ 218 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. ELSQWVNMEEFKSHSPERSIFSAIWEGNCHFEQHQGQEEGYFRQLMINHENMPIFSQHTLLTQEFYDREKISECKKCRKIFSYHLFFSHHKRTHSKEL | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Sandwich ELISA | |
| 4G11 | |
| Western Blot 1:500, ELISA, Sandwich ELISA | |
| NP_694989 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 92283 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction