missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOLT1B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA564276
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: RRVPVLGSLL NLPGIRSFVD KVGESN Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
GOT1B may be involved in fusion of ER-derived transport vesicles with the Golgi complex.Specifications
GOLT1B | |
Polyclonal | |
Unconjugated | |
GOLT1B | |
2310061A22Rik; AA407874; AU024291; CGI-141; EGK_03451; GCT2; Germ cell tumor 2; Golgi transport 1 homolog B; golgi transport 1 homolog B (S. cerevisiae); golgi transport 1B; Golgi transport 1-like protein B; GOLT1B; GOT1; GOT1A; GOT1B; HDCMA39P; hGOT1a; putative NF-kappa-B-activating protein 470; RGD1310691; UNQ432/PRO793; Vesicle transport protein GOT1B; YMR292W | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51026 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y3E0 | |
GOLT1B | |
Recombinant protein corresponding to Human GOLT1B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only