Learn More
Goat IgG VisUCyte HRP Polymer, R&D Systems™
GREENER_CHOICE
Product Code. 18782819
Change view
Click to view available options
Quantity:
125 mL
25 mL
50 mL (2 x 25 mL)
Unit Size:
125mL
25mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18782819 25 mL 25mL
18783819 50 mL (2 x 25 mL) 50mL
18730319 125 mL 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18782819 Supplier R&D Systems Supplier No. VC004025
Please to purchase this item. Need a web account? Register with us today!
Donkey Polyclonal Antibody has been used in 2 publications
IgG Polyclonal specifically detects IgG in Goat samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Antigen IgG
Applications Immunohistochemistry
Classification Polyclonal
Conjugate HRP Polymer
Dilution Immunohistochemistry
Formulation Supplied as a solution in PBS with preservative.
Gene Alias Immunoglobulin G, ImmunoglobulinG
Host Species Donkey
Immunogen Goat IgG
Purification Method Affinity Purified
Quantity 25 mL
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Test Specificity Detects heavy ( gamma ) chains on goat IgG and light chains on all goat immunoglobulins, based on IEP. No cross-reactivity is observed with non-immunoglobulin goat serum proteins or serum proteins from human, mouse or rat, based on IEP. VisUCyte™ HRP Polymer provides high sensitivity with low background in IHC.
Content And Storage 6 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG