missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Monkey IgM Heavy Chain Secondary Antibody, Alkaline Phosphatase, Novus Biologicals™

Product Code. 18262023 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. Quantity unitSize
18262023 1 mg 1mg
1 options
This item is not returnable. View return policy

Product Code. 18262023

Brand: Novus Biologicals NBP172664

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

IgM Heavy Chain Polyclonal antibody specifically detects IgM Heavy Chain in Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgM Heavy Chain
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Alkaline Phosphatase (AP)
Dilution Western Blot 1:500 - 1:3000, ELISA 1:2000 - 1:10000, Immunohistochemistry 1:200 - 1:1000
Gene Alias Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain
Gene Symbols IGHM
Host Species Goat
Immunogen Monkey IgM u chain
Purification Method Affinity Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3507
Test Specificity No reaction was observed against other Monkey heavy or light chain proteins.
Target Species Monkey
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.