Learn More
Goat anti-Human IgG Fc Secondary Antibody (Pre-adsorbed), Novus Biologicals™
Goat Polyclonal Antibody
Brand:  Novus Biologicals NBP1-74995-0.5mg
 View more versions of this product
Product Code. 18538553
  • 137.00 € / 0.50mg
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoassay, Immunomicroscopy, Chemiluminescence Immunoassay.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG Fc
Polyclonal
Unconjugated
Goat
Affinity purified
RUO
Based on IEP, this antibody reacts with heavy gamma chains on human IgG. This antibody has been pre-adsorbed against human IgA + IgM.
Store at 4°C. After dilution, add an equal volume of glycerol to prevent loss of enzymatic activity and store at -20°C. Avoid repeated freeze/thaw cycles.
Immunoassay
4.5 mg/mL
O75015, O75015, O75015
Purified Human IgG, Fc fragment
0.5 mg
Secondary