missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Horse IgG whole molecule, Biotin, Polyclonal, Secondary Antibody, Abnova™
Brand: Abnova PAB10647.2mg
This item is not returnable.
View return policy
Specifications
| Goat anti-Horse IgG whole molecule | |
| Polyclonal | |
| Goat anti-horse IgG recognizes horse IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Biotin. | |
| Lyophilized from 10mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| Horse IgG whole molecule | |
| 2 mg | |
| Equine | |
| Lyophilized |
| ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
| Biotin | |
| The optimal working dilution should be determined by the end user. | |
| Goat | |
| This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. | |
| Secondary | |
| Store at 4°C on dry atmosphere. After reconstitution with 1 mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
Hazard Statement
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : 5-chloro-2-methyl-3-isothiazolone
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction